CAS RN 144-33-2
IUPAC Navn:
disodium 3-carboxy-3-hydroxypentanedioate
Synonymer:
disodium citrate
Molekylvægt (g/mol):
236.09
Molekylformel:
C6H6Na2O7
InChi Key:
CEYULKASIQJZGP-UHFFFAOYSA-L
SMILES:
[Na+].[Na+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
1
–
3
af
3
Resultater
1
Disodium Citrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Disodium Citrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Disodium Citrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.