CAS RN 14220-21-4
IUPAC Navn:
bromorhenium; pentakis(methanidylidyneoxidanium)
Synonymer:
bromopentacarbonylrhenium(I)
Molekylvægt (g/mol):
406.16
Molekylformel:
C5BrO5Re
InChi Key:
NWJBOTGGBYFKEJ-UHFFFAOYSA-M
SMILES:
Br[Re].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
1
–
4
af
4
Resultater
2
Bromopentacarbonylrhenium(I), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Bromopentacarbonylrhenium(I), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Bromopentacarbonylrhenium(I), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.