CAS RN 142-18-7
IUPAC Navn:
2,3-dihydroxypropyl dodecanoate
Synonymer:
1-monomyristin C12-18 acid triglyceride monolaurin
Molekylvægt (g/mol):
274.4
Molekylformel:
C15H30O4
InChi Key:
ARIWANIATODDMH-UHFFFAOYNA-N
SMILES:
CCCCCCCCCCCC(=O)OCC(O)CO
1
–
3
af
3
Resultater
1
Monolaurin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Monolaurin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Monolaurin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.