Få mere at vide
Monolaurin, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Monolaurin |
| CAS | 142-18-7 |
| Molekylær formel | C15H30O4 |
| Mængde | 100 mg |
| Synonym | 1-Mono-laurin, (±)-2,3-Dihydroxypropyl dodecanoate, (±)-Glyceryl 1-monododecanoate, 1-Glyceryl laurate, 1-Monododecanoylglycerol, 1-Monolaurin, 1-Monolauroyl-rac-glycerol, 3-Dodecanoyloxy-1,2-propanediol, Dodecanoic acid alpha-monoglyceride, Glycerin 1-monolaurate, Glycerol 1-laurate, Glycerol 1-monododecanoate, Glycerol 1-monolaurate, Glycerol alpha-monolaurate, Glyceryl laurate, Glyceryl monododecanoate, Glyceryl monolaurate, Lauric acid 1-monoglyceride, Lauric acid alpha-monoglyceride, Lauricidin, Luaricidin, NSC 698570, alpha-Monolaurin |
| InChI formel | InChI=1S/C15H30O4/c1-2-3-4-5-6-7-8-9-10-11-15(18)19-13-14(17)12-16/h14,16-17H,2-13H2,1H3 |
| SMIL | CCCCCCCCCCCC(=O)OCC(O)CO |
| IUPAC navn | 2,3-dihydroxypropyl dodecanoate |
| Molekylvægt (g/mol) | 274.4 |
| Formel vægt | 274.2144 |