CAS RN 141109-15-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(S)-(+)-2-Chlorophenylglycine Methyl Ester Tartrate Salt, TRC
CAS: 141109-15-1 Molekylær formel: C9 H10 Cl N O2 . C4 H6 O6 Molekylvægt (g/mol): 349.72 Synonym: Benzeneacetic acid, α-amino-2-chloro-, methyl ester, (αS)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1),Benzeneacetic acid, α-amino-2-chloro-, methyl ester, (+)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) IUPAC navn: (2R,3R)-2,3-dihydroxybutanedioic acid;methyl (2S)-2-amino-2-(2-chlorophenyl)acetate SMIL: COC(=O)[C@@H](N)c1ccccc1Cl.O[C@H]([C@@H](O)C(=O)O)C(=O)O
2
Methyl (2S)-2-Amino-2-(2-chlorophenyl)acetate Tartrate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.