CAS RN 1402438-74-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Tocris Bioscience™ PF 06273340
Potent and selective pan-Trk inhibitor; peripherally restricted
2