Få mere at vide
Tocris Bioscience™ PF 06273340, Tocris Bioscience™
GREENER_CHOICE
Potent and selective pan-Trk inhibitor; peripherally restricted
Brand:  Tocris Bioscience™ 6115/10
Encompass
Produktkode 15747226
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
PF 06273340 is a potent and selective pan-Trk inhibitor (IC50 values are 1, 2 and 6 nM for Trk C, B and A, respectively). Selective for Trk over a panel of ion channels, receptors and other enzymes. Peripherally restricted. Exhibits efficacy in a rodent model of inflammatory pain. Orally bioavailable.
Tekniske data
Tekniske data
PF 06273340
N-[5-[[2-Amino-7-(2-hydroxy-1,1-dimethylethyl)-7H-pyrrolo[2,3-d]pyrimidin-5-yl]carbonyl]-3-pyridinyl]-5-chloro-2-pyridineacetamide
Soluble to 100mM in DMSO
Orange
1402438-74-7
10 mg
Solid
Bioactive Small Molecule
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only.