CAS RN 1398065-84-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
(±)-Quizalofop-ethyl-d3 (propionate-3,3,3-d3), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
(±)-Quizalofop-ethyl-d3 (propionate-3,3,3-d3), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(±)-Quizalofop-ethyl-d3 (propionate-3,3,3-d3), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
(±)-Quizalofop-ethyl-d3 (propionate-3,3,3-d3), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.