Få mere at vide
(±)-Quizalofop-ethyl-d3 (propionate-3,3,3-d3), TRC
Artikelnummer. 30419294
Skift visning
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
Artikelnummer. Mængde unitSize
30419294 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419294 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ8261090.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Quizalofop-ethylD3(3,3,3-D3)
CAS 1398065-84-3
Renhedsgrad noter HPLC
Molekylær formel C19 2H3 H14 Cl N2 O4
Mængde 0.5 mg
Synonym Quizalofop-ethyl D3 (3,3,3 D3), Quizalofop Ethyl D3 (methyl D3), Propanoic-3,3,3-d3 acid','2-[4-[(6-chloro-2-quinoxalinyl)oxy]phenoxy]-','ethyl ester (ACI), Ethyl 2-[4-[(6-chloro-2-quinoxalinyl)oxy]phenoxy]propanoate-3,3,3-d3 (ACI), Quizalofop-p-ethyl-d3
InChI formel InChI=1S/C19H17ClN2O4/c1-3-24-19(23)12(2)25-14-5-7-15(8-6-14)26-18-11-21-17-10-13(20)4-9-16(17)22-18/h4-12H,3H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])C(Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1)C(=O)OCC
IUPAC navn ethyl 2-[4-(6-chloroquinoxalin-2-yl)oxyphenoxy]-3,3,3-trideuteriopropanoate
Molekylvægt (g/mol) 375.82
Formel vægt 375.1065
Procent renhed >95