CAS RN 135-97-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
psi-Tropine, TRC
CAS: 135-97-7 Molekylær formel: C8 H15 N O Molekylvægt (g/mol): 141.21 Synonym: Pseudotropine,3β-Tropanol,1αH,5αH-Tropan-3β-ol (8CI),8-Azabicyclo[3.2.1]octan-3-ol, 8-methyl-, exo-,Pseudotropine (6CI,7CI),(3-exo)-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol,3-Pseudotropanol,3β-Tropanol,NSC 43871,Pseudotropanol,ψ-Tropine IUPAC navn: (1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol SMIL: CN1[C@@H]2CC[C@H]1C[C@H](O)C2
2
Pseudotropine 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality