CAS RN 1346604-70-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Candesartan-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Candesartan-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Candesartan-d4, MedChemExpress
MedChemExpress Candesartan D4 (CV-11974 D4) is the deuterium labeled Candesartan, which is an angiotensin II receptor antagonist.