Få mere at vide
Candesartan-d4, TRC
Artikelnummer. 30342290
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30342290 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342290 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17557810MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Candesartan-d4
CAS 1346604-70-3
Renhedsgrad noter HPLC
Molekylær formel C24 2H4 H16 N6 O3
Mængde 10 mg
Synonym 2-Ethoxy-1-[[2'-(1H-tetrazol-5-yl)[1-(phenyl-d4)-1’-phenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic Acid, CV-11974-d4, Candesartan M1-d4, 3-[[2'-(1H-Tetrazol-5-yl)[1-(phenyl-d4)-1’-phenyl]-4-yl]methyl]-2-ethoxy-3H-benzimidazole-4-carboxylic Acid
InChI formel InChI=1S/C24H20N6O3/c1-2-33-24-25-20-9-5-8-19(23(31)32)21(20)30(24)14-15-10-12-16(13-11-15)17-6-3-4-7-18(17)22-26-28-29-27-22/h3-13H,2,14H2,1H3,(H,31,32)(H,26,27,28,29)/i10D,11D,12D,13D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1Cn2c(OCC)nc3cccc(C(=O)O)c23)c4ccccc4c5nn[nH]n5
IUPAC navn 2-ethoxy-3-[[2,3,5,6-tetradeuterio-4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylic acid
Molekylvægt (g/mol) 444.48
Formel vægt 444.1848
Procent renhed >95