CAS RN 1346601-19-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
4-(2-Hydroxy-3-isopropylaminopropoxy)benzoic Acid 2-Hydroxyethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Hydroxyethyl 4-[((2RS)-2-Hydroxy-3-(isopropylamino)propyl)oxy]benzoate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
4-(2-Hydroxy-3-isopropylaminopropoxy)benzoic Acid 2-Hydroxyethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.