CAS RN 13453-80-0
IUPAC Navn:
lithium(1+) dihydrogen phosphate
Synonymer:
lithium(1+) dihydrogen phosphate
Molekylvægt (g/mol):
103.93
Molekylformel:
H2LiO4P
InChi Key:
SNKMVYBWZDHJHE-UHFFFAOYSA-M
SMILES:
[Li+].OP(O)([O-])=O