CAS RN 1330165-98-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Acebutolol Dimer Impurity, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N,N'-[[(1-Methylethyl)imino]-bis[(2-hydroxypropane-1,3-diyl)oxy(3-acetyl-1,4-phenylene)]]dibutanamide (Acebutolol Biamine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.