CAS RN 132810-10-7
IUPAC Navn:
1-ethyl-4-[4-(4-fluorophenyl)-5H,6H,7H,8H,9H,10H-cycloocta[b]pyridin-2-yl]piperazine
Synonymer:
blonanserin
Molekylvægt (g/mol):
367.51
Molekylformel:
C23H30FN3
InChi Key:
XVGOZDAJGBALKS-UHFFFAOYSA-N
SMILES:
CCN1CCN(CC1)C1=CC(C2=CC=C(F)C=C2)=C2CCCCCCC2=N1
1
–
3
af
3
Resultater
1
Blonanserin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Blonanserin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Blonanserin, MedChemExpress
MedChemExpress Blonanserin (AD-5423) is a potent and orally active 5-HT2A (Ki=0.812 nM) and dopamine D2 receptor (Ki =0.142 nM) antagonist. Blonanserin is usually acts as an atypical antipsychotic agent and can be used for the research of extrapyramidal symptoms, excessive sedation, or hypotension.