CAS RN 132-66-1
IUPAC Navn:
2-[(naphthalen-1-yl)carbamoyl]benzoic acid
Synonymer:
naptalam N-1-napthylphthalamic acid N-1-naphthyl-phthalamidsaeure NPA α-naphthylphthalamic acid npa (naptalam)
Molekylvægt (g/mol):
291.31
Molekylformel:
C18H13NO3
InChi Key:
JXTHEWSKYLZVJC-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC=C1C(=O)NC1=C2C=CC=CC2=CC=C1
1
–
3
af
3
Resultater
1
N-(1-Naphthyl)phthalamic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-(1-Naphthyl)phthalamic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-(1-Naphthyl)phthalamic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.