CAS RN 1313738-90-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
PGMI-004A, MedChemExpress
MedChemExpress PGMI-004A is a potent phosphoglycerate mutase 1 (PGAM1) inhibitor with an IC50 of 13.1 μM.