CAS RN 13113-71-8
IUPAC Navn:
2-hydroxy-2-phenylpropanoic acid
Synonymer:
(+-)-atrolactic acid (1)-2-phenyllactic acid hydroxymethylmandelic acid atrolactic acid, (+-)- atrolactinic acid dl-atrolactic acid (+-)-isomer atrolactic acid dl-atrolactate (S)-atrolactic acid atrolactinate
Molekylvægt (g/mol):
166.18
Molekylformel:
C9H10O3
InChi Key:
NWCHELUCVWSRRS-UHFFFAOYNA-N
SMILES:
CC(O)(C(O)=O)C1=CC=CC=C1
1
–
1
af
1
Resultater
1
(S)-(+)-2-Hydroxy-2-phenylpropionic acid, 98+%
CAS: 13113-71-8 Molekylær formel: C9H10O3 Molekylvægt (g/mol): 166.18 MDL nummer: MFCD00067700 InChI nøgle: NWCHELUCVWSRRS-UHFFFAOYNA-N Synonym: s-atrolactic acid,2s-2-hydroxy-2-phenylpropanoic acid,s-+-2-hydroxy-2-phenylpropionic acid,atrolactic acid, +,s-2-hydroxy-2-phenylpropanoic acid,unii-22y6g519ru,s-+-atrolactic acid,atrolactic acid 2-phenyl-lactic acid,benzeneacetic acid, a-hydroxy-a-methyl-, as PubChem CID: 445144 ChEBI: CHEBI:40741 IUPAC navn: (2S)-2-hydroxy-2-phenylpropansyre SMIL: CC(O)(C(O)=O)C1=CC=CC=C1