CAS RN 131061-50-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(3Alpha,5Alpha,11Beta)-3,11,17-trihydroxy-21-[(tetrahydro-2H-pyran-2-yl)oxy]-pregnan-20-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3Alpha,5Alpha,11Beta)-3,11,17-trihydroxy-21-[(tetrahydro-2H-pyran-2-yl)oxy]-pregnan-20-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3a,5a,11b)-3,11,17-trihydroxy-21-[(tetrahydro-2H-pyran-2-yl)oxy]-pregnan-20-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.