CAS RN 130683-47-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Methyl 2-Chloro-4-fluoro-5-(2,6-dioxo-3-methyl-4-trifluoromethylpyrimidin-1-yl)benzoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl 2-Chloro-4-fluoro-5-(2,6-dioxo-3-methyl-4-trifluoromethylpyrimidin-1-yl)benzoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl 2-Chloro-4-fluoro-5-(2,6-dioxo-3-methyl-4-trifluoromethylpyrimidin-1-yl)benzoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.