CAS RN 1292821-90-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
K-80003, MedChemExpress
MedChemExpress K-80003 is a potent inhibitor of tRXRα-dependent Akt activation and cancer cell growth.