CAS RN 1286770-55-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
5
af
5
Resultater
1
Verubecestat, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Verubecestat, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Tocris Bioscience™ Verubecestat
BACE1 and BACE2 inhibitor
4
Verubecestat, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Verubecestat, MedChemExpress
MedChemExpress Verubecestat (MK-8931) is an orally active, high-affinity BACE1 and BACE2 inhibitor with Ki values of 2.2 nM and 0.38 nM. Verubecestat effectively reduces Aβ40 and has the potential for Alzheimer's Disease.