Få mere at vide
Verubecestat, MedChemExpress
Artikelnummer. 16443207
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16443207 50 mg 50mg
16427176 100 mg -
16423546 25 mg -
16419717 5 mg -
16417188 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16443207 Leverandør Medchem Express Leverandørnr. HY16759
MedChemExpress Verubecestat (MK-8931) is an orally active, high-affinity BACE1 and BACE2 inhibitor with Ki values of 2.2 nM and 0.38 nM. Verubecestat effectively reduces Aβ40 and has the potential for Alzheimer's Disease.
Verubecestat (MK-8931) is an orally active, high-affinity BACE1 and BACE2 inhibitor with Ki values of 2.2 nM and 0.38 nM. Verubecestat effectively reduces Aβ40 and has the potential for Alzheimer's Disease.
Tekniske data
Kemisk navn eller materiale Verubecestat
CAS 1286770-55-5
Farve White
Renhedsgrad noter Research
Molekylær formel C17H17F2N5O3S
Mængde 50 mg
Synonym MK-8931
Opløselighedsinformation DMSO : ≥ 35 mg/mL (85.49 mM)
SMIL O=C(C1=NC=C(F)C=C1)NC2=CC=C(F)C([C@@](C3)(C)N=C(N)N(C)S3(=O)=O)=C2
Molekylvægt (g/mol) 409.41
Formel vægt 409.41
Procent renhed 98.98%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neurodegeneration