CAS RN 128008-87-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2-Nitrofluorene-d9, TRC
CAS: 128008-87-7 Molekylær formel: C13 D9 N O2 Molekylvægt (g/mol): 220.2715 Synonym: 9H-Fluorene-1,2,3,4,5,6,8,9,9-d9, 7-nitro- (9CI),7-Nitro-9H-fluorene-1,2,3,4,5,6,8,9,9-d9,2-Nitrofluorene-1,3,4,5,6,7,8,9,9-d9,2-Nitrofluorene-d9,2-Nitrofluorene-D9,2-Nitro-9H-fluorene-d9 IUPAC navn: 1,2,3,4,5,6,8,9,9-nonadeuterio-7-nitrofluorene SMIL: [2H]c1c([2H])c([2H])c2c(c1[2H])c3c([2H])c([2H])c(c([2H])c3C2([2H])[2H])[N+](=O)[O-]
2
2-Nitrofluorene-d9, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.