Få mere at vide
2-Nitrofluorene-d9, TRC
Artikelnummer. 30481996
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481996 10 mg
30464335 1 mg
30467162 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481996 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49384210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 128008-87-7
Molekylær formel C13 D9 N O2
Molekylvægt (g/mol) 220.2715
Synonym 9H-Fluorene-1,2,3,4,5,6,8,9,9-d9, 7-nitro- (9CI), 7-Nitro-9H-fluorene-1,2,3,4,5,6,8,9,9-d9, 2-Nitrofluorene-1,3,4,5,6,7,8,9,9-d9, 2-Nitrofluorene-d9, 2-Nitrofluorene-D9, 2-Nitro-9H-fluorene-d9
IUPAC navn 1,2,3,4,5,6,8,9,9-nonadeuterio-7-nitrofluorene
SMIL [2H]c1c([2H])c([2H])c2c(c1[2H])c3c([2H])c([2H])c(c([2H])c3C2([2H])[2H])[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 2-Nitrofluorene (D9)
Mængde 10 mg
InChI formel InChI=1S/C13H9NO2/c15-14(16)11-5-6-13-10(8-11)7-9-3-1-2-4-12(9)13/h1-6,8H,7H2/i1D,2D,3D,4D,5D,6D,7D2,8D
Formel vægt 220.1198