Få mere at vide
2-Nitrofluorene-d9, CDN
Artikelnummer. 30324827
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324827 0.1 g 0.1g
30318780 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324827 Leverandør CDN Leverandørnr. CDND52330.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Nitrofluorene (D9)
CAS 128008-87-7
Molekylær formel C13 D9 N O2
Mængde 0.1 g
Synonym 9 H-Fluorene-1,2,3,4,5,6,8,9,9-d9, 7-nitro- (9 CI), 7-Nitro-9 H-fluorene-1,2,3,4,5,6,8,9,9-d9, 2-Nitrofluorene-1,3,4,5,6,7,8,9,9-d9, 2-Nitrofluorene-d9, 2-Nitrofluorene-D9, 2-Nitro-9 H-fluorene-d9
InChI formel InChI=1 S/C13H9NO2/c15-14(16)11-5-6-13-10(8-11)7-9-3-1-2-4-12(9)13/h1-6,8 H,7H2/i1D,2 D,3 D,4 D,5 D,6 D,7D2,8 D
SMIL [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c3c([2 H])c([2 H])c(c([2 H])c3C2([2 H])[2 H])[N+](=O)[O-]
IUPAC navn 1,2,3,4,5,6,8,9,9-nonadeuterio-7-nitrofluorene
Molekylvægt (g/mol) 220.2715
Formel vægt 220.1198 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity