CAS RN 126860-83-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(1Alpha,3Beta,5Z,7E,22E)-24-Cyclopropyl-1,3-dihydroxy-9,10-Secochola-5,7,10(19),22-tetraen-24-one (90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
MC 1046, MedChemExpress
MedChemExpress MC 1046(Impurity A of Calcipotriol) is an impurity of Calcipotriol; Calcipotriol (MC 903; Calcipotriene) is a ligand of VDR-like receptors.