CAS RN 1262417-51-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
NMS-P118, MedChemExpress
MedChemExpress NMS-P118 is a potent, orally available, and highly selective PARP-1 Inhibitor for cancer therapy.