CAS RN 126-75-0
IUPAC Navn:
diethyl {[2-(ethylsulfanyl)ethyl]sulfanyl}phosphonate
Synonymer:
demeton demeton-S
Molekylvægt (g/mol):
258.33
Molekylformel:
C8H19O3PS2
InChi Key:
GRPRVIYRYGLIJU-UHFFFAOYSA-N
SMILES:
CCOP(=O)(OCC)SCCSCC
1
–
3
af
3
Resultater
1
Demeton-S 1000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality