CAS RN 1252637-35-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Petesicatib, MedChemExpress
MedChemExpress Petesicatib is a cathepsin S inhibitor, used in research of immune diseases.