CAS RN 1246819-17-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Nitazoxanide-d4, TRC
CAS: 1246819-17-9 Molekylær formel: C12H5D4N3O5S Molekylvægt (g/mol): 311.31 Synonym: 2-(Acetyloxy)-N-(5-nitro-2-thiazolyl)benzamide-d4,Alinia-d4,BRN 1225475-d4,Cryptaz-d4,DRG 0242-d4,N-(5-Nitro-2-thiazolyl)salicylamide-d4 Acetate Ester,Nitacure-d4,Nitarid-d4,Nitazoxanida-d4,Nixide-d4,Nixoran-d4,Nizonide-d4,PH 5776-d4 SMIL: [2H]c1c([2H])c([2H])c(C(=O)Nc2ncc(s2)[N+](=O)[O-])c(OC(=O)C)c1[2H]
2
Nitazoxanide-d4, MedChemExpress
MedChemExpress Nitazoxanide D4 (NTZ D4) is the deuterium labeled Nitazoxanide, which is an antiprotozoal agent.