Få mere at vide
Nitazoxanide-d4, TRC
Artikelnummer. 30478532
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30478532 1 mg
30477472 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30478532 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4901021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246819-17-9
Molekylær formel C12H5D4N3O5S
Molekylvægt (g/mol) 311.31
Synonym 2-(Acetyloxy)-N-(5-nitro-2-thiazolyl)benzamide-d4, Alinia-d4, BRN 1225475-d4, Cryptaz-d4, DRG 0242-d4, N-(5-Nitro-2-thiazolyl)salicylamide-d4 Acetate Ester, Nitacure-d4, Nitarid-d4, Nitazoxanida-d4, Nixide-d4, Nixoran-d4, Nizonide-d4, PH 5776-d4
SMIL [2H]c1c([2H])c([2H])c(C(=O)Nc2ncc(s2)[N+](=O)[O-])c(OC(=O)C)c1[2H]
Tekniske data
Kemisk navn eller materiale Nitazoxanide-d4
Mængde 1 mg
InChI formel InChI=1S/C12H9N3O5S/c1-7(16)20-9-5-3-2-4-8(9)11(17)14-12-13-6-10(21-12)15(18)19/h2-6H,1H3,(H,13,14,17)/i2D,3D,4D,5D