CAS RN 1246086-78-1
IUPAC Navn:
N-{3-chloro-4-[(2-chlorophenyl)sulfamoyl]phenyl}pyridine-2-carboxamide
Synonymer:
N-{3-chloro-4-[(2-chlorophenyl)sulfamoyl]phenyl}pyridine-2-carboxamide
Molekylvægt (g/mol):
422.28
Molekylformel:
C18H13Cl2N3O3S
InChi Key:
IXHCGJXBIHHIEF-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC=C1NS(=O)(=O)C1=C(Cl)C=C(NC(=O)C2=CC=CC=N2)C=C1
1
–
1
af
1
Resultater
1
VU 0364439, MedChemExpress
MedChemExpress VU 0364439 is a mGlu4 positive allosteric modulator (PAM), with EC50 of 19.8 nM.