CAS RN 124379-29-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(4S)-(3',4'-Dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(4S)-4-(3,4-dichlorophenyl)-3,4-dihydronaphthalen-1(2H)-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
(4S)-(3',4'-Dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.