CAS RN 123798-55-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Sodium Nitrite-18O2 (10% 16O2), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Sodium Nitrite-18O2 may contain up to 2% of 16O2, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Sodium Nitrite-18O2 may contain up to 2% of 16O2, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.