missing translation for 'onlineSavingsMsg'
Få mere at vide

Sodium Nitrite-18O2 (10% 16O2), TRC

Artikelnummer. 30417285
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417285 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30417285 Leverandør Toronto Research Chemicals Leverandørnr. TRCS64800310MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Sodium Nitrite-18O2
CAS 123798-55-0
Molekylær formel N 18O2 . Na
Mængde 10 mg
Synonym Nitrous-18O2 Acid Sodium Salt (10% 16O2)
InChI formel InChI=1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1/i2+2,3+2;
SMIL [Na+].[18O-]N=[18O]
Molekylvægt (g/mol) 72.995
Formel vægt 72.991
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.