CAS RN 1234703-40-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Globalagliatin, MedChemExpress
MedChemExpress Globalagliatin (LY2608204) is a activator of glucokinase (GK) with EC50 of 42 nM.