CAS RN 123086-68-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Δ7-Naltrexone (7,8-Didehydronaltrexone; (5alpha)-17-(Cyclopropylmethyl)-7,8-didehydro-4,5-epoxy-3,14-dihydroxymorphinan-6-one), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
7,8-Didehydronaltrexone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.