CAS RN 1228817-38-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Mps1-IN-2, MedChemExpress
MedChemExpress Mps1-IN-2 is a potent, selective and ATP-competitive dual Mps1/Plk1 inhibitor, with an IC50 and a Kd of 145 nM and 12 nM for Mps1 and a Kd of 61 nM for Plk1.