Få mere at vide
Mps1-IN-2, MedChemExpress
Artikelnummer. 16476386
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16476386 100 mg -
16465958 10 mg -
16452107 50 mg -
16428587 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16476386 Leverandør Medchem Express Leverandørnr. HY13994
MedChemExpress Mps1-IN-2 is a potent, selective and ATP-competitive dual Mps1/Plk1 inhibitor, with an IC50 and a Kd of 145 nM and 12 nM for Mps1 and a Kd of 61 nM for Plk1.
Mps1-IN-2 is a potent, selective and ATP-competitive dual Mps1/Plk1 inhibitor, with an IC50 and a Kd of 145 nM and 12 nM for Mps1 and a Kd of 61 nM for Plk1.
Tekniske data
Kemisk navn eller materiale Mps1-IN-2
CAS 1228817-38-6
Farve Light Brown
Renhedsgrad noter Research
Molekylær formel C26H36N6O3
Mængde 100 mg
Opløselighedsinformation DMSO : 13.33 mg/mL (27.74 mM; Need ultrasonic)
SMIL CN(C1=CN=C(NC2=C(OCC)C=C(N3CCC(O)CC3)C=C2)N=C1N(C4CCCC4)CC5)C5=O
Molekylvægt (g/mol) 480.6
Formel vægt 480.6
Procent renhed 98.15%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease