CAS RN 1224510-29-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
beta-Cypermethrin, TRC
CAS: 1224510-29-5 Molekylær formel: C22 H19 Cl2 N O3 Molekylvægt (g/mol): 416.3 Synonym: Beta-cypermethrin,Chinmix,Cypermethrin-beta,Cypermethrin-β,β-Cypermethrin IUPAC navn: [(R)-cyano-(3-phenoxyphenyl)methyl] (1S)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate SMIL: CC1(C)C(C=C(Cl)Cl)[C@@H]1C(=O)O[C@@H](C#N)c2cccc(Oc3ccccc3)c2
2
beta-Cypermetrin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality