Få mere at vide
beta-Cypermethrin, TRC
Artikelnummer. 30367931
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367931 1 mg
30358360 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30367931 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9893901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1224510-29-5
Molekylær formel C22 H19 Cl2 N O3
Molekylvægt (g/mol) 416.3
Synonym Beta-cypermethrin, Chinmix, Cypermethrin-beta, Cypermethrin-β, β-Cypermethrin
IUPAC navn [(R)-cyano-(3-phenoxyphenyl)methyl] (1S)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
SMIL CC1(C)C(C=C(Cl)Cl)[C@@H]1C(=O)O[C@@H](C#N)c2cccc(Oc3ccccc3)c2
Tekniske data
Kemisk navn eller materiale beta-Cypermethrin
Mængde 1 mg
InChI formel InChI=1S/C22H19Cl2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/t17?,18-,20+/m0/s1
Formel vægt 415.0742