CAS RN 1222780-33-7
IUPAC Navn:
3-(1-propanoylpiperidin-4-yl)-1-[4-(trifluoromethoxy)phenyl]urea
Synonymer:
3-(1-propanoylpiperidin-4-yl)-1-[4-(trifluoromethoxy)phenyl]urea
Molekylvægt (g/mol):
359.35
Molekylformel:
C16H20F3N3O3
InChi Key:
AAJMQTLFRTZCJK-UHFFFAOYSA-N
SMILES:
CCC(=O)N1CCC(CC1)NC(=O)NC1=CC=C(OC(F)(F)F)C=C1
1
–
1
af
1
Resultater
1
TPPU, MedChemExpress
MedChemExpress TPPU is a soluble epoxide hydrolase (sEH) inhibitor with IC50 values of 37 and 3.7 nM for monkey and human sEH, respectively.