CAS RN 1219804-53-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2-Methyl-d3-propyl-2,3,3,3-d4 Alcohol, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
2-Methyl-1-propanol-d7, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.