Få mere at vide
2-Methyl-1-propanol-d7, TRC
Artikelnummer. 30408282
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30408282 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408282 Leverandør Toronto Research Chemicals Leverandørnr. TRCI78007850MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyl-1-propanol-d7
CAS 1219804-53-1
Molekylær formel C4 D7 H3 O
Mængde 50 mg
Synonym 1-Propan-2,3,3,3-d4-ol, 2-(methyl-d3)- (ACI), 2-(Methyl-d3)-1-propan-2,3,3,3-d4-ol (ACI), 2-Methyl-d3-propyl-2,3,3,3-d4 alcohol, Isobutanol-d7, Isobutyl-d7 alcohol, 2-Methyl-1-propanol-d7, 2-Methyl-1-propanol-D7 (isopropyl-D7)
InChI formel InChI=1S/C4H10O/c1-4(2)3-5/h4-5H,3H2,1-2H3/i1D3,2D3,4D
SMIL [2H]C([2H])([2H])C([2H])(CO)C([2H])([2H])[2H]
IUPAC navn 2,3,3,3-tetradeuterio-2-(trideuteriomethyl)propan-1-ol
Molekylvægt (g/mol) 81.165
Formel vægt 81.117