Få mere at vide
2-Methyl-d3-propyl-2,3,3,3-d4 Alcohol, CDN
Artikelnummer. 30312515
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30312515 0.25 g
30309271 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312515 Leverandør CDN Leverandørnr. CDND57390.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyl-1-propanol-d7
CAS 1219804-53-1
Molekylær formel C4 D7 H3 O
Mængde 0.25 g
Synonym 1-Propan-2,3,3,3-d4-ol, 2-(methyl-d3)- (ACI), 2-(Methyl-d3)-1-propan-2,3,3,3-d4-ol (ACI), 2-Methyl-d3-propyl-2,3,3,3-d4 alcohol, Isobutanol-d7, Isobutyl-d7 alcohol, 2-Methyl-1-propanol-d7, 2-Methyl-1-propanol-D7 (isopropyl-D7)
InChI formel InChI=1 S/C4H10O/c1-4(2)3-5/h4-5 H,3H2,1-2H3/i1D3,2D3,4 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(CO)C([2 H])([2 H])[2 H]
IUPAC navn 2,3,3,3-tetradeuterio-2-(trideuteriomethyl)propan-1-ol
Molekylvægt (g/mol) 81.165
Formel vægt 81.117 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity