CAS RN 1214265-58-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
WZ4003, MedChemExpress
MedChemExpress WZ4003 is the first potent and highly specific NUAK kinase inhibitor with IC50 of 20 nM/100 nM for NUAK1 (ARK5)/NUAK2, without significant inhibition on other 139 kinases.