Få mere at vide
WZ4003, MedChemExpress
Artikelnummer. 16469387
Encompass_Preferred
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16469387 5 mg
16422917 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16469387 Leverandør Medchem Express Leverandørnr. HY15802
MedChemExpress WZ4003 is the first potent and highly specific NUAK kinase inhibitor with IC50 of 20 nM/100 nM for NUAK1 (ARK5)/NUAK2, without significant inhibition on other 139 kinases.
WZ4003 is the first potent and highly specific NUAK kinase inhibitor with IC50 of 20 nM/100 nM for NUAK1 (ARK5)/NUAK2, without significant inhibition on other 139 kinases.
Tekniske data
Kemisk navn eller materiale WZ4003
CAS 1214265-58-3
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C25H29ClN6O3
Mængde 5 mg
Opløselighedsinformation DMSO : 33.33 mg/mL (67.06 mM; Need ultrasonic)
SMIL O=C(NC1=CC=CC(OC2=NC(NC3=C(OC)C=C(N4CCN(C)CC4)C=C3)=NC=C2Cl)=C1)CC
Molekylvægt (g/mol) 496.99
Formel vægt 496.99
Procent renhed 98.88%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease