CAS RN 1207989-09-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
LGD-6972, MedChemExpress
MedChemExpress LGD-6972 is a selective and orally active glucagon receptor antagonist. LGD-6972 has the potential for type 2 diabetes research.